4-(4-Chlorophenoxy)aniline

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Amino-4'-chlorodiphenyl ether

009744-10G 009744-50G
FLUO009744-10GEA 0 GBP
FLUO009744-10G FLUO009744-50G
4-(4-Chlorophenoxy)aniline
4-(4-Chlorophenoxy)aniline
Formula: C₁₂H₁₀ClNO
MW: 219.67 g/mol
MDL Number: MFCD00043925
CAS Number: 101-79-1
EINECS: 202-976-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR