(4-Acetamidophenyl)boronic acid pinacol ester

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

010823-5G 010823-1G
FLUO010823-5GEA 0 GBP
FLUO010823-5G FLUO010823-1G
(4-Acetamidophenyl)boronic acid pinacol ester
(4-Acetamidophenyl)boronic acid pinacol ester
Formula: C₁₄H₂₀BNO₃
MDL Number: MFCD02093722
CAS Number: 214360-60-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR