(3-Acetamidophenyl)boronic acid pinacol ester
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
010822-5G
010822-1G
FLUO010822-5GEA
0
GBP
FLUO010822-5G
FLUO010822-1G
(3-Acetamidophenyl)boronic acid pinacol ester
(3-Acetamidophenyl)boronic acid pinacol ester
Formula:
C₁₄H₂₀BNO₃ |
MDL Number:
MFCD03789261 CAS Number: 480424-93-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...