(2-Acetamidophenyl)boronic acid pinacol ester

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

010821-5G 010821-1G
FLUO010821-5GEA 0 GBP
FLUO010821-5G FLUO010821-1G
(2-Acetamidophenyl)boronic acid pinacol ester
(2-Acetamidophenyl)boronic acid pinacol ester
Formula: C₁₄H₂₀BNO₃
MDL Number: MFCD02179480
CAS Number: 380430-61-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR