Methyl-4-bromocrotonate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Methyl 4-bromobut-2-enoate

M-Clarity™ quality level = MQ100

M32100-100G M32100-25G M32100-5G
SIALM32100-100GEA 410 GBP
SIALM32100-100G SIALM32100-25G SIALM32100-5G
Methyl-4-bromocrotonate, Sigma-Aldrich®
Methyl 4-bromocrotonate
Formula: C₅H₇BrO₂
MW: 179.01 g/mol
MDL Number: MFCD00000246
CAS Number: 1117-71-1
EINECS: 214-251-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR