Progesterone, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ200

P3972-25G P3972-5G
SIALP3972-25GEA 282 GBP
SIALP3972-25G SIALP3972-5G
Progesterone, Sigma-Aldrich®
Progesterone
Formula: C₂₁H₃₀O₂
MDL Number: MFCD00003658
CAS Number: 57-83-0
EINECS: 200-350-6

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR