Progesterone, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ200
P0130-25G
P0130-100G
SIALP0130-25GEA
182
GBP
SIALP0130-25G
SIALP0130-100G
Progesterone, Sigma-Aldrich®
Progesterone
Formula:
C₂₁H₃₀O₂ |
MDL Number:
MFCD00003658 CAS Number: 57-83-0 EINECS: 200-350-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...