5-Thiazolecarboxylic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

016173-1G 016173-5G
FLUO016173-1GEA 0 GBP
FLUO016173-1G FLUO016173-5G
5-Thiazolecarboxylic acid
5-Thiazolecarboxylic acid
Formula: C₄H₃NO₂S
MW: 129.1371 g/mol
MDL Number: MFCD03428539
CAS Number: 14527-41-4
EINECS: 238-545-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR