6-Bromo-3-coumaranone
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
6-Bromobenzofuran-3(2H)-one
AS2698G1
AS2698G5
ACTIAS2698G1EA
0
GBP
ACTIAS2698G1
ACTIAS2698G5
6-Bromo-3-coumaranone
6-Bromo-3-coumaranone
Formula:
C₈H₅BrO₂ MW: 213.0281 g/mol |
MDL Number:
MFCD08437617 CAS Number: 201809-69-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...