2-Methylfuran-3-boronic acid pinacol ester

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4,4,5,5-Tetramethyl-2-(2-methyl-3-furyl)-1,3,2-dioxaborolane

PN-9091-1G PN-9091-250M
COBBPN-9091-1GEA 0 GBP
COBBPN-9091-1G COBBPN-9091-250M
2-Methylfuran-3-boronic acid pinacol ester
2-Methylfuran-3-boronic acid pinacol ester
Formula: C₁₁H₁₇BO₃
MDL Number: MFCD09879905
CAS Number: 864776-02-3
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR