3-Formylfuran-2-boronic acid
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
(3-Formylfuran-2-yl)boronic acid
011037-5G
011037-1G
FLUO011037-5GEA
0
GBP
FLUO011037-5G
FLUO011037-1G
3-Formylfuran-2-boronic acid
3-Formylfuran-2-boronic acid
Formula:
C₅H₅BO₄ MW: 139.90 g/mol |
MDL Number:
MFCD01075680 CAS Number: 27339-38-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...