Furan-2-boronic acid pinacol ester

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-(2-Furanyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-Furanboronic acid pinacol ester

PN-2309-1G PN-2309-5G
COBBPN-2309-1GEA 0 GBP
COBBPN-2309-1G COBBPN-2309-5G
Furan-2-boronic acid pinacol ester
Furan-2-boronic acid pinacol ester
Formula: C₁₀H₁₅BO₃
MW: 194.04 g/mol
MDL Number: MFCD02094008
CAS Number: 374790-93-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR