3,4-(Methylenedioxy)phenylacetonitrile
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
002596-5G
002596-25G
FLUO002596-5GEA
0
GBP
FLUO002596-5G
FLUO002596-25G
3,4-(Methylenedioxy)phenylacetonitrile
3,4-(Methylenedioxy)phenylacetonitrile
Formula:
C₉H₇NO₂ |
MDL Number:
MFCD00005835 CAS Number: 4439-02-5 EINECS: 224-655-9 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...