5-Iodo-1,3-benzodioxole
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
5-Iodobenzo[d][1,3]dioxole
018746-5G
018746-25G
FLUO018746-5GEA
0
GBP
FLUO018746-5G
FLUO018746-25G
5-Iodo-1,3-benzodioxole
5-Iodo-1,3-benzodioxole
Formula:
C₇H₅IO₂ MW: 248.0179 g/mol |
MDL Number:
MFCD00079766 CAS Number: 5876-51-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...