Triethyl-2-fluoro-2-phosphonoacetate (fluorinated horner reagent)

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Diethyl [(ethoxycarbonyl)(fluoro)methyl]phosphonate, Triethyl 2-fluoro-2-phosphonoacetate

006355-5G 006355-10G 006355-25G
FLUO006355-5GEA 0 GBP
FLUO006355-5G FLUO006355-10G FLUO006355-25G
Triethyl-2-fluoro-2-phosphonoacetate (fluorinated horner reagent)
Triethyl 2-fluoro-2-phosphonoacetate (fluorinated Horner reagent)
Formula: C₈H₁₆FO₅P
MDL Number: MFCD00134400
CAS Number: 2356-16-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR