4-(Aminomethyl)piperidine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

011323-25G 011323-100G
FLUO011323-25GEA 0 GBP
FLUO011323-25G FLUO011323-100G
4-(Aminomethyl)piperidine
4-(Aminomethyl)piperidine
Formula: C₆H₁₄N₂
MW: 114.1888 g/mol
MDL Number: MFCD00006007
CAS Number: 7144-05-0
EINECS: 230-446-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR