Trimethyl phosphonoacetate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Methyl 2-(dimethoxyphosphoryl)acetate, Dimethyl methoxycarbonylmethanephosphonate

M-Clarity™ quality level = MQ200

T79758-100G T79758-25G
SIALT79758-100GEA 138 GBP
SIALT79758-100G SIALT79758-25G
Trimethyl phosphonoacetate, Sigma-Aldrich®
Trimethyl phosphonoacetate
Formula: C₅H₁₁O₅P
MW: 182.11 g/mol
MDL Number: MFCD00008452
CAS Number: 5927-18-4
EINECS: 227-663-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR