6-Chloro-8-(trifluoromethyl)quinoline

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-6999-5G HC-6999-1G
COBBHC-6999-5GEA 0 GBP
COBBHC-6999-5G COBBHC-6999-1G
6-Chloro-8-(trifluoromethyl)quinoline
6-Chloro-8-(trifluoromethyl)quinoline
Formula: C₁₀H₅ClF₃N
MDL Number: MFCD11053881
CAS Number: 1065074-68-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR