4-Chloro-8-(trifluoromethyl)quinoline

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-6338-5G HC-6338-1G
COBBHC-6338-5GEA 0 GBP
COBBHC-6338-5G COBBHC-6338-1G
4-Chloro-8-(trifluoromethyl)quinoline
4-Chloro-8-(trifluoromethyl)quinoline
Formula: C₁₀H₅ClF₃N
MDL Number: MFCD00134578
CAS Number: 23779-97-7
EINECS: 245-880-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR