4-Chloro-7-(trifluoromethyl)quinoline

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CFF992.1 CFF992.10 CFF992.25 004561-5G 004561-1G
CHMPCFF992.1EA 0 GBP
CHMPCFF992.1 CHMPCFF992.10 CHMPCFF992.25 FLUO004561-5G FLUO004561-1G
4-Chloro-7-(trifluoromethyl)quinoline
4-Chloro-7-(trifluoromethyl)quinoline
Formula: C₁₀H₅ClF₃N
MDL Number: MFCD00006775
CAS Number: 346-55-4
EINECS: 206-471-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR