4-Chloro-2-(trifluoromethyl)quinoline

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

004559-1G 004559-5G
FLUO004559-1GEA 0 GBP
FLUO004559-1G FLUO004559-5G
4-Chloro-2-(trifluoromethyl)quinoline
4-Chloro-2-(trifluoromethyl)quinoline
Formula: C₁₀H₅ClF₃N
MDL Number: MFCD00153105
CAS Number: 1701-24-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR