4-Chloro-2-(trifluoromethyl)quinoline
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HC-2174-5G
HC-2174-25G
HC-2174-1G
COBBHC-2174-5GEA
0
GBP
COBBHC-2174-5G
COBBHC-2174-25G
COBBHC-2174-1G
4-Chloro-2-(trifluoromethyl)quinoline
4-Chloro-2-(trifluoromethyl)quinoline
Formula:
C₁₀H₅ClF₃N |
MDL Number:
MFCD00153105 CAS Number: 1701-24-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...