4,8-Dichloro-2-(trifluoromethyl)quinoline

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CFF253.2.5 008282-1G 008282-5G
CHMPCFF253.2.5EA 0 GBP
CHMPCFF253.2.5 FLUO008282-1G FLUO008282-5G
4,8-Dichloro-2-(trifluoromethyl)quinoline
4,8-Dichloro-2-(trifluoromethyl)quinoline
Formula: C₁₀H₄Cl₂F₃N
MDL Number: MFCD00276590
CAS Number: 18706-35-9

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR