1,4-Bis(trifluoromethyl)-2-nitrobenzene
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2-Nitro-1,4-bis(trifluoromethyl)benzene, 1-Nitro-2,5-bis(trifluoromethyl)benzene
006030-1G
006030-5G
FLUO006030-1GEA
0
GBP
FLUO006030-1G
FLUO006030-5G
1,4-Bis(trifluoromethyl)-2-nitrobenzene
1,4-Bis(trifluoromethyl)-2-nitrobenzene
Formula:
C₈H₃F₆NO₂ MW: 259.11 g/mol |
MDL Number:
MFCD00236674 CAS Number: 320-88-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...