1-Chloro-2,4-bis(trifluoromethyl)benzene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1,3-Bis(trifluoromethyl)-4-chlorobenzene

005570-5G 005570-1G
FLUO005570-5GEA 0 GBP
FLUO005570-5G FLUO005570-1G
1-Chloro-2,4-bis(trifluoromethyl)benzene
1-Chloro-2,4-bis(trifluoromethyl)benzene
Formula: C₈H₃ClF₆
MW: 248.56 g/mol
MDL Number: MFCD01631638
CAS Number: 327-76-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR