4-(Chlorodifluoromethyl)benzotrifluoride

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

010270-1G 010270-5G
FLUO010270-1GEA 0 GBP
FLUO010270-1G FLUO010270-5G
4-(Chlorodifluoromethyl)benzotrifluoride
4-(Chlorodifluoromethyl)benzotrifluoride
Formula: C₈H₄ClF₅
CAS Number: 13947-94-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR