1-Bromo-2,4-bis(trifluoromethyl)benzene

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2,4-Bis(trifluoromethyl)-1-bromobenzene

OS-7565-5G OS-7565-25G OS-7565-100G
COBBOS-7565-5GEA 0 GBP
COBBOS-7565-5G COBBOS-7565-25G COBBOS-7565-100G
1-Bromo-2,4-bis(trifluoromethyl)benzene
1-Bromo-2,4-bis(trifluoromethyl)benzene
Formula: C₈H₃BrF₆
MW: 293.01 g/mol
MDL Number: MFCD00074904
CAS Number: 327-75-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR