4-Bromo-2-nitro-5-(trifluoromethyl)aniline
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
5-Amino-2-bromo-4-nitrobenzotrifluoride
HC-2840-1G
HC-2840-5G
COBBHC-2840-1GEA
0
GBP
COBBHC-2840-1G
COBBHC-2840-5G
4-Bromo-2-nitro-5-(trifluoromethyl)aniline
4-Bromo-2-nitro-5-(trifluoromethyl)aniline
Formula:
C₇H₄BrF₃N₂O₂ MW: 285.0181 g/mol |
MDL Number:
MFCD09263997 CAS Number: 683241-86-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...