n-Butyl phenyl ether

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: N-butylphenylether

001072-100G 001072.500G 001072-25ML
FLUO001072-100GEA 0 GBP
FLUO001072-100G FLUO001072.500G FLUO001072-25ML
n-Butyl phenyl ether
n-Butyl phenyl ether
Formula: C₁₀H₁₄O
MW: 150.22 g/mol
MDL Number: MFCD00009438
CAS Number: 1126-79-0
EINECS: 214-426-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR