(S)-(+)-Nipecotic acid
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AM-1713-5G
AM-1713-25G
AM-1713-1G
COBBAM-1713-5GEA
0
GBP
COBBAM-1713-5G
COBBAM-1713-25G
COBBAM-1713-1G
(S)-(+)-Nipecotic acid
(S)-(+)-Nipecotic acid
Formula:
C₆H₁₁NO₂ |
MDL Number:
MFCD01630807 CAS Number: 59045-82-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...