(R)-(-)-Nipecotic acid
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
(R)-Piperidine-3-carboxylic acid, (R)-(-)-3-Piperidinecarboxylic acid, D-Nipecotic acid
AM-1714-5G
AM-1714-25G
AM-1714-1G
COBBAM-1714-5GEA
0
GBP
COBBAM-1714-5G
COBBAM-1714-25G
COBBAM-1714-1G
(R)-(-)-Nipecotic acid
(R)-(-)-Nipecotic acid
Formula:
C₆H₁₁NO₂ MW: 129.16 g/mol |
MDL Number:
MFCD01630787 CAS Number: 25137-00-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...