Ferrocenecarbaldehyde, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Ferrocenecarboxaldehyde

MQ100

122459-25G 122459-5G
SIAL122459-25GEA 206 GBP
SIAL122459-25G SIAL122459-5G
Ferrocenecarbaldehyde, Sigma-Aldrich®
Ferrocenecarbaldehyde
Formula: C₁₁H₁₀FeO
MW: 214.04 g/mol
MDL Number: MFCD00001429
CAS Number: 12093-10-6
EINECS: 235-158-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR