2,4-Difluoro-5-(trifluoromethyl)aniline

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 5-Amino-2,4-difluorobenzotrifluoride

013827-5G 013827-25G
FLUO013827-5GEA 0 GBP
FLUO013827-5G FLUO013827-25G
2,4-Difluoro-5-(trifluoromethyl)aniline
2,4-Difluoro-5-(trifluoromethyl)aniline
Formula: C₇H₄F₅N
MDL Number: MFCD01631613
CAS Number: 261944-56-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR