(3-Fluoro-4-methylphenyl)acetic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-(3-Fluoro-4-methylphenyl)acetic acid

005805-1G 005805-2.5G
FLUO005805-1GEA 0 GBP
FLUO005805-1G FLUO005805-2.5G
(3-Fluoro-4-methylphenyl)acetic acid
(3-Fluoro-4-methylphenyl)acetic acid
Formula: C₉H₉FO₂
MDL Number: MFCD01631536
CAS Number: 261951-74-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR