3,4-Dimethoxy-β-nitrostyrene
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
018588-10G
018588-25G
FLUO018588-10GEA
0
GBP
FLUO018588-10G
FLUO018588-25G
3,4-Dimethoxy-β-nitrostyrene
3,4-Dimethoxy-β-nitrostyrene
Formula:
C₁₀H₁₁NO₄ |
MDL Number:
MFCD00024823 CAS Number: 4230-93-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...