α-Hydroxy-α-methylbenzyl phenyl ketone ≥98%
Supplier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2-Hydroxy-1,2-diphenylpropan-1-one
L07240.09
L07240.18
L07240.09EA
0
GBP
L07240.09
L07240.18
α-Hydroxy-α-methylbenzyl phenyl ketone ≥98%
α-Hydroxy-α-methylbenzyl phenyl ketone
Purity: 98%
CAS No.: 5623-26-7
Molecular Formula: C15H14O2
Formula Weight: 226.28
Melting Point: 63-67°
Beilstein Reference: 2049961
Formula:
C₁₅H₁₄O₂ Melting Pt: 63…67 °C |
MDL Number:
MFCD00051804 CAS Number: 5623-26-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...