1,1,2,3,3,3-Hexafluoropropyl dichloromethyl ether
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
010319-1G
010319-5G
010319-25G
FLUO010319-1GEA
0
GBP
FLUO010319-1G
FLUO010319-5G
FLUO010319-25G
1,1,2,3,3,3-Hexafluoropropyl dichloromethyl ether
1,1,2,3,3,3-Hexafluoropropyl dichloromethyl ether
Formula:
C₄H₂Cl₂F₆O |
MDL Number:
MFCD03094547 CAS Number: 56860-82-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...