1,1,2,3,3,3-Hexafluoropropyl chlorofluoromethyl ether

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

010318-1G 010318-5G
FLUO010318-1GEA 0 GBP
FLUO010318-1G FLUO010318-5G
1,1,2,3,3,3-Hexafluoropropyl chlorofluoromethyl ether
1,1,2,3,3,3-Hexafluoropropyl chlorofluoromethyl ether
Formula: C₄H₂ClF₇O
MDL Number: MFCD03094548
CAS Number: 56860-86-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR