α,α-Difluorophenylacetic acid
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
α,α-Difluorobenzeneacetic acid
HC-2559-5G
HC-2559-1G
COBBHC-2559-5GEA
0
GBP
COBBHC-2559-5G
COBBHC-2559-1G
α,α-Difluorophenylacetic acid
α,α-Difluorophenylacetic acid
Formula:
C₈H₆F₂O₂ MW: 172.13 g/mol |
MDL Number:
MFCD00498146 CAS Number: 360-03-2 UN: 3261 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...