3-(Trifluoromethoxy)toluene
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OS-7538-5G
OS-7538-25G
OS-7538-1G
COBBOS-7538-5GEA
0
GBP
COBBOS-7538-5G
COBBOS-7538-25G
COBBOS-7538-1G
3-(Trifluoromethoxy)toluene
3-(Trifluoromethoxy)toluene
Formula:
C₈H₇F₃O |
MDL Number:
MFCD07366526 CAS Number: 705-44-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...