3-Fluoro-5-methoxyphenylboronic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-Fluoro-5-methoxybenzeneboronic acid

016207-1G 016207-5G
FLUO016207-1GEA 0 GBP
FLUO016207-1G FLUO016207-5G
3-Fluoro-5-methoxyphenylboronic acid
3-Fluoro-5-methoxyphenylboronic acid
Formula: C₇H₈BFO₃
MW: 169.95 g/mol
MDL Number: MFCD07363780
CAS Number: 609807-25-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR