2-Fluoro-3-(trifluoromethyl)benzyl alcohol

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

006409-1G 006409-5G 006409-10G
FLUO006409-1GEA 0 GBP
FLUO006409-1G FLUO006409-5G FLUO006409-10G
2-Fluoro-3-(trifluoromethyl)benzyl alcohol
2-Fluoro-3-(trifluoromethyl)benzyl alcohol
Formula: C₈H₆F₄O
MW: 194.13 g/mol
MDL Number: MFCD00061166
CAS Number: 207981-45-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR