3-Bromophenylboronic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

016107-1G 016107-5G
FLUO016107-1GEA 0 GBP
FLUO016107-1G FLUO016107-5G
3-Bromophenylboronic acid
3-Bromophenylboronic acid
Formula: C₆H₆BBrO₂
MDL Number: MFCD00239386
CAS Number: 89598-96-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR