3-Chlorobenzylamine ≥98.0% (by GC, titration analysis)

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: (3-Chlorophenyl)methanamine

C1089-25G C1089-5G
TCIAC1089-25GEA 0 GBP
TCIAC1089-25G TCIAC1089-5G
3-Chlorobenzylamine ≥98.0% (by GC, titration analysis)
3-Chlorobenzylamine
Formula: C₇H₈ClN
MW: 141.6 g/mol
Boiling Pt: 226°C
Density: 1,159 g/cm³ (20 °C)
Flash Pt: 98°C
MDL Number: MFCD00040752
CAS Number: 4152-90-3
EINECS: 223-985-0
UN: 2735
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7ecb54b55f885a7b1ff50e43e2d20496efa69881&vid=7ecb54b55f885a7b1ff50e43e2d20496efa69881&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR