Flash chromatography columns, PuriFlash® FM, ALB
Supplier: INTERCHIM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
FM70ALB/20G FM70ALB/10G FM15ALB/2G FM25ALB/5G FM150ALB/70G FM150ALB/50G FM150ALB/25G
INTEFM70ALB/20GEA 243 GBP
INTEFM70ALB/20G INTEFM70ALB/10G INTEFM15ALB/2G INTEFM25ALB/5G INTEFM150ALB/70G INTEFM150ALB/50G INTEFM150ALB/25G
Flash chromatography columns, PuriFlash® FM, ALB
Chromatography Columns
Columns for basic compound purification. Eliminate the use of TEA (triethylamine) to avoid streaking or tailing under normal or reverse phase conditions.
Order Now
SPECIFICATIONS
Environmentally Preferable