2-(Aminomethyl)-15-crown-5, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ100
388416-1G
388416-250MG
SIAL388416-1GEA
263
GBP
SIAL388416-1G
SIAL388416-250MG
2-(Aminomethyl)-15-crown-5, Sigma-Aldrich®
2-(Aminomethyl)-15-crown-5
Formula:
C₁₁H₂₃NO₅ |
CAS Number:
83585-56-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...