Coumalic acid, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2-Oxo-2H-pyran-5-carboxylic acid
M-Clarity™ quality level = MQ100
C85409-25G
C85409-5G
SIALC85409-25GEA
119
GBP
SIALC85409-25G
SIALC85409-5G
Coumalic acid, Sigma-Aldrich®
Coumalic acid
Formula:
C₆H₄O₄ MW: 140.10 g/mol |
MDL Number:
MFCD00006644 CAS Number: 500-05-0 EINECS: 207-899-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...