α,α-Diphenyl-N-methyl-L-prolinol ≥97%, ee 99%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

L19399.03 L19399.MD
L19399.03EA 0 GBP
L19399.03 L19399.MD
α,α-Diphenyl-N-methyl-L-prolinol ≥97%, ee 99%
α,α-Diphenyl-N-methyl-L-prolinol

Purity: 97% CAS No.: 110529-22-1 Molecular Formula: C18H21NO Formula Weight: 267.37 Storage Temperature: 2°C to 8°C Melting Point: 66-69° Beilstein Reference: 4749387 Optical Rotation: +52° (c=1 in chloroform) MDL No.: MFCD00145245

Formula: C₁₈H₂₁NO
Melting Pt: 66…69 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00145245
CAS Number: 110529-22-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR