Dicyclohexano-18-crown-6 (mixture of isomers)

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

002918-5G 002918-1G
FLUO002918-5GEA 0 GBP
FLUO002918-5G FLUO002918-1G
Dicyclohexano-18-crown-6 (mixture of isomers)
Dicyclohexano-18-crown-6 (mixture of isomers)
Formula: C₂₀H₃₆O₆
MDL Number: MFCD00005099
CAS Number: 16069-36-6
EINECS: 240-216-4
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR