5-Acetylthiophene-2-boronic acid ≥98%
Supplier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
L15221.03
L15221.06
L15221.14
L15221.03EA
19.2
GBP
L15221.03
L15221.06
L15221.14
5-Acetylthiophene-2-boronic acid ≥98%
5-Acetylthiophene-2-boronic acid
Purity: 98%
CAS No.: 206551-43-1
Molecular Formula: C6H7BO3S
Formula Weight: 170.00
Melting Point: 142-148°
Beilstein Reference: 8681302
Formula:
C₆H₇BO₃S Melting Pt: 142…148 °C Storage Temperature: Ambient |
MDL Number:
MFCD01075681 CAS Number: 206551-43-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...